1-Chlorobutane 99.5% for analysis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: n-Butyl Chloride

44702-0010
TS44702-0010EA 89 USD
TS44702-0010
1-Chlorobutane 99.5% for analysis
1-Chlorobutane
Formula: CH₃(CH₂)₃Cl
MW: 92.57 g/mol
Boiling Pt: 77…78 °C (760 mmHg)
Melting Pt: –123 °C
Density: 0.88 g/cm³
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00001009
CAS Number: 109-69-3
UN: 1127
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2bd980eb6e7ecd91951d46d0ea510119e976dc0&vid=5231549172a609ad96623a80204b11c7c0c69273&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4010 to 1.4030 (20 °C, 589 nm)
Water ≤0.05 %
GC ≥99.4 %
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Acidity ≤0.0005 meq/g
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Zinc (Zn) ≤0.1 ppm
Sulfur compounds (as S) ≤0.002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR