Lithium-tert-butanolate 8% (w/w) 0.9 M in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC44684-1000 AC44684-8000
TS44684-1000EA 65 USD
TS44684-1000 TS44684-8000
Lithium-tert-butanolate 8% (w/w) 0.9 M in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal™
Lithium tert-butanolate
Formula: C₄H₉LiO
Density: 0.86 g/cm³
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00050479
CAS Number: 1907-33-1
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear yellow to brown Solution (May develop some turbidity or precipitate)
Strength 0.80 to 1.10 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR