Magnesium nitrate hexahydrate for biochemistry

Supplier: Thermo Scientific Chemicals

44683-0025 44683-5000
TS44683-0025EA 284 USD
TS44683-0025 TS44683-5000
Magnesium nitrate hexahydrate for biochemistry
Magnesium nitrate hexahydrate
Formula: Mg(NO₃)₂·6H₂O
Boiling Pt: 330 °C (760 mmHg)
Melting Pt: 89 °C
Flash Pt: 93.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00149779
CAS Number: 13446-18-9

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Titration Complexometric 98 to 102 %
pH 5.0 to 8.2 (at 25°C)
Chloride (Cl) ≤0.001 %
Phosphate (PO4) ≤0.0005 %
Sulfate (SO4) ≤0.005 %
Ammonium (NH4) ≤0.003 %
Barium (Ba) ≤0.005 %
Calcium (Ca) ≤0.01 %
Iron (Fe) ≤0.0005 %
Manganese (Mn) ≤0.0005 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.005 %
Strontium (Sr) ≤0.005 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR