Sodium acetate trihydrate 99+% for biochemistry

Supplier: Thermo Scientific Chemicals

AC44679-0010 AC44679-2500
TS44679-0010EA 70 USD
TS44679-0010 TS44679-2500
Sodium acetate trihydrate 99+% for biochemistry
Sodium acetate trihydrate
Formula: H₃CCOONa·3H₂O
Melting Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00071557
CAS Number: 6131-90-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystals
Infrared spectrum Conforms
UV at 280 nm A: ≤0.05
pH 7.5 to 9.5
Titration with HClO4 99.0 to 101.0 %
Chloride (Cl) ≤0.001 %
Phosphate (PO4) ≤0.0005 %
Sulfate (SO4) ≤0.002 %
Calcium (Ca) ≤0.0005 %
Iron (Fe) ≤0.0005 %
Magnesium (Mg) ≤0.0005 %
Potassium (K) ≤0.005 %
Zinc (Zn) ≤0.0005 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR