Potassium nitrate 99%, pure

Supplier: Thermo Scientific Chemicals
Warning

AC44637-0025 AC44637-0100 AC44637-5000
TS44637-0025EA 140 USD
TS44637-0025 TS44637-0100 TS44637-5000
Potassium nitrate 99%, pure
Potassium nitrate
Formula: KNO₃
Boiling Pt: 400 °C (760 mmHg)
Melting Pt: 334 °C
Storage Temperature: Ambient
MDL Number: MFCD00011409
CAS Number: 7757-79-1
UN: 1486
ADR: 5.1,III

Order Now

Specification Test Results

Appearance White Crystalline powder and/or chunks
Loss on drying ≤0.5 %
Titration after Ion exchange ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR