Sodium dihydrogen phosphate dihydrate for HPLC

Supplier: Thermo Scientific Chemicals

Synonyms: Phosphoric acid (sodium hydrate), Sodium phosphate monobasic dihydrate, Sodium dihydrogen orthophosphate dihydrate

AC44622-2500
TS44622-2500EA 66 USD
TS44622-2500
Sodium dihydrogen phosphate dihydrate for HPLC
Sodium dihydrogen phosphate dihydrate
Formula: NaH₂PO₄·2H₂O
MW: 156.01 g/mol
Melting Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00149209
CAS Number: 13472-35-0
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 12,08806

Order Now

Specification Test Results

Appearance (Color) White
Appearance (Form) Crystals
UV at 254 nm A (1 M in water) =<0.05
pH 4.0 to 4.5
Titration Acid 98.5 to 101.5 %
Water =<24 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR