Sodium acetate trihydrate for HPLC
Supplier: Thermo Scientific Chemicals
Synonyms:
Acetic acid sodium salt trihydrate
AC44620-0010
AC44620-2500
TS44620-0010EA
125
USD
TS44620-0010
TS44620-2500
Sodium acetate trihydrate for HPLC
Sodium acetate trihydrate
Formula:
H₃CCOONa·3H₂O Melting Pt: 58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00071557 CAS Number: 6131-90-4 |
|
Specification Test Results
Appearance | White Crystals |
UV | at 254 nm A: ≤0.02 |
pH | 7.5 to 9.2 |
Titration with HClO4 | ≥98.5 % |
Insoluble matter | ≤0.0005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...