Sodium acetate trihydrate for HPLC

Supplier: Thermo Scientific Chemicals

Synonyms: Acetic acid sodium salt trihydrate

AC44620-0010 AC44620-2500
TS44620-0010EA 125 USD
TS44620-0010 TS44620-2500
Sodium acetate trihydrate for HPLC
Sodium acetate trihydrate
Formula: H₃CCOONa·3H₂O
Melting Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00071557
CAS Number: 6131-90-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystals
UV at 254 nm A: ≤0.02
pH 7.5 to 9.2
Titration with HClO4 ≥98.5 %
Insoluble matter ≤0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR