Pyrrolidine 99.5+%, AcroSeal™, purified by redistillation

Supplier: Thermo Scientific Chemicals
Danger

44614-1000 44614-8000
TS44614-1000EA 68 USD
TS44614-1000 TS44614-8000
Pyrrolidine 99.5+%, AcroSeal™, purified by redistillation
Pyrrolidine
Formula: C₄H₉N
Boiling Pt: 86…88 °C (760 mmHg)
Melting Pt: –63 °C
Density: 0.866 g/cm³
Flash Pt: 3 °C
Storage Temperature: Ambient
MDL Number: MFCD00005249
CAS Number: 123-75-1
UN: 1922
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4421 to 1.4441 (20°C, 589 nm)
Water ≤0.2 %
GC ≥99.4 %
Color scale ≤10 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR