EDTA tetrasodium salt dihydrate 99%

Supplier: Thermo Scientific Chemicals
Danger

44608-0010 44608-1000 44608-5000
TS44608-0010EA 138 USD
TS44608-0010 TS44608-1000 TS44608-5000
EDTA tetrasodium salt dihydrate 99%
EDTA tetrasodium salt dihydrate
Formula: C₁₀H₁₂N₂Na₄O₈·2H₂O
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150026
CAS Number: 10378-23-1

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Complexometric 99.0 to 102.0 % (on dry substance)
Loss on drying 6.4 % to 10.6 %
Solubility (16% in water) Clear solution


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR