Hydrogen chloride 3 M in CPME (cyclopentyl methyl ether), AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

44585-1000
TS44585-1000EA 94 USD
TS44585-1000
Hydrogen chloride 3 M in CPME (cyclopentyl methyl ether), AcroSeal™
Hydrogen chloride
Formula: HCl
Boiling Pt: 106 °C
Density: 0.895 g/cm³
Flash Pt: –1 °C
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 2920
ADR: 8,I

Order Now

Specification Test Results

Appearance Colorless to light yellow Solution
Strength 2.85 to 3.30 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR