N-Methyl-2-pyrrolidone (NMP) 99.5% for analysis
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, N-Methylpyrrolidinone, N-Methyl-2-pyrrolidinone, N-Methyl-alpha-pyrrolidone, M-Pyrol, N-methylpyrrolidone, N-Methyl-gamma-butyrolactam, 1-Methyl-2-pyrrolidone, NMP, N-Methylpyrrolidone, N-Methyl-2-pyrrolidone, Methyl Pyrrolidone
44579-0010
445790025
TS44579-0010EA
129
USD
TS44579-0010
TS445790025
N-Methyl-2-pyrrolidone (NMP) 99.5% for analysis
N-Methyl-2-pyrrolidone (NMP)
Formula:
C₅H₉NO MW: 99.13 g/mol Boiling Pt: 202 Melting Pt: -24 Density: 1.03 g/cm³ Flash Pt: 91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003193 CAS Number: 872-50-4 Merck Index: 13,06140 |
|
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Water | ≤0.1 % |
GC | ≥99.4 % |
Color scale | ≤50 APHA |
Residue after ignition | ≤0.01 % |
Aluminium (Al) | ≤1 ppm |
Antimony (Sb) | ≤1 ppm |
Arsenic (As) | ≤1 ppm |
Boron (B) | ≤1 ppm |
Calcium (Ca) | ≤5 ppm |
Chromium (Cr) | ≤1 ppm |
Copper (Cu) | ≤1 ppm |
Iron (Fe) | ≤1 ppm |
Lead (Pb) | ≤1 ppm |
Magnesium (Mg) | ≤1 ppm |
Manganese (Mn) | ≤1 ppm |
Nickel (Ni) | ≤1 ppm |
Potassium (K) | ≤20 ppm |
Sodium (Na) | ≤20 ppm |
Zinc (Zn) | ≤1 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...