2-Ethoxyethanol for analysis

Supplier: Thermo Scientific Chemicals
Danger

445760010 445760025
TS445760010EA 86 USD
TS445760010 TS445760025
2-Ethoxyethanol for analysis
2-Ethoxyethanol
Formula: C₂H₅OCH₂CH₂OH
Boiling Pt: 135 °C (760 mmHg)
Melting Pt: –70 °C
Density: 0.93 g/cm³
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00002869
CAS Number: 110-80-5
UN: 1171
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.1 %
GC ≥98.5 %
Color scale ≤10 APHA
Residue after evaporation ≤0.005 %
Acidity/alkalinity ≤0.001 meq/g
Calcium (Ca) ≤1 ppm
Copper (Cu) ≤0.05 ppm
Iron (Fe) ≤0.5 ppm
Lead (Pb) ≤0.05 ppm
Magnesium (Mg) ≤0.2 ppm
Potassium (K) ≤0.5 ppm
Silicon (Si) ≤0.05 ppm
Sodium (Na) ≤2 ppm
Sulfur (S) ≤2 ppm
Zinc (Zn) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR