Tetrachloroethylene 99+% for HPLC

Supplier: Thermo Scientific Chemicals
Warning

445690010 445690025 445691000
TS445690010EA 147 USD
TS445690010 TS445690025 TS445691000
Tetrachloroethylene 99+% for HPLC
Tetrachloroethylene
Formula: C₂Cl₄
Boiling Pt: 120…122 °C (760 mmHg)
Melting Pt: –22 °C
Density: 1.625 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000834
CAS Number: 127-18-4
UN: 1897
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.05 %
GC ≥99.0 %
UV at 400 nm A: ≤0.03
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR