Methanol, Technical Grade

Supplier: Thermo Scientific Chemicals
Danger

44431-0050
TS44431-0050EA 54 USD
TS44431-0050
Methanol, Technical Grade
Methanol
Formula: CH₃OH
Boiling Pt: 64.7 °C (760 mmHg)
Melting Pt: –98 °C
Density: 0.791 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
UN: 1230
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Refractive index 1.3280 to 1.3300 (20°C, 589 nm)
Water ≤0.5 %
GC ≥99 %
Ethanol ≤0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR