Heptane (mixture of isomers), Technical Grade

Supplier: Thermo Scientific Chemicals
Danger

44427-0050
TS44427-0050EA 97 USD
TS44427-0050
Heptane (mixture of isomers), Technical Grade
Heptane (mixture of isomers)
Formula: H₃C(CH₂)₅CH₃
MW: 100.2 g/mol
Boiling Pt: 98 °C (760 mmHg)
Melting Pt: –91 °C
Density: 0.683 g/cm³
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
UN: 1206
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dabde8793eb27ef796098b842bf18b9ced8539b8&vid=d30a8083280d1915a43f51d93c49f9b76eabd579&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Boiling point 94 to 100°C
Density 0.685 to 0.730 kg/l (15°C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR