Xylene (mixture of isomers), Technical Grade

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dimethylbenzene

444240050
TS444240050EA 45 USD
TS444240050
Xylene (mixture of isomers), Technical Grade
Xylene (mixture of isomers)
Formula: C₆H₄(CH₃)₂
MW: 106.17 g/mol
Boiling Pt: 136…140 °C (760 mmHg)
Melting Pt: –34 °C
Density: 0.865 g/cm³
Flash Pt: 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00077264
CAS Number: 1330-20-7
UN: 1307
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Density 0.860 to 0.875 kg/l
GC ≥98.5 % (Mixture of isomers)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR