Paraffin, liquid, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Wax Paraffin, Paraffin wax

444180250
TS444180250EA 262 USD
TS444180250
Paraffin, liquid, Technical Grade
Paraffin
Boiling Pt: 300 °C (760 mmHg)
Melting Pt: –24 °C
Density: 0.88 g/cm³
Flash Pt: 218 °C
Storage Temperature: Ambient
CAS Number: 8002-74-2

Order Now

Specification Test Results

Appearance Clear colorless Oily liquid
Infrared spectrum Conforms
Refractive index 1.4600 to 1.4800 (20°C, 589 nm)
Density (20°C) 0.810 to 0.890 g/ml
Viscosity 20 to 80 mm²/s (40°C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR