Picolinamide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Pyridinecarboxamide, Pyridine-2-carboxamide

44336-0100 44336-0500
TS44336-0100EA 99 USD
TS44336-0100 TS44336-0500
Picolinamide 98%
Picolinamide
Formula: C₆H₆N₂O
MW: 122.13 g/mol
Boiling Pt: 143 °C (2.7kPa)
Melting Pt: 106…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00023483
CAS Number: 1452-77-3

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR