1-Bromo-4-ethynylbenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

44279-0010
TS44279-0010EA 100 USD
TS44279-0010
1-Bromo-4-ethynylbenzene 98%
1-Bromo-4-ethynylbenzene
Formula: C₈H₅Br
Boiling Pt: 71…72 °C (3Torr)
Melting Pt: 64…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00168822
CAS Number: 766-96-1
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder or solid
Infrared spectrum Conforms
Melting point 62 to 68 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR