Strontium nitrate 98%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

AC43852-0010 AC43852-0050
TS43852-0010EA 88 USD
TS43852-0010 TS43852-0050
Strontium nitrate 98%, Extra Pure
Strontium nitrate
Formula: Sr(NO₃)₂
Boiling Pt: 645 °C (1013 hPa)
Melting Pt: 570 °C
Storage Temperature: Ambient
MDL Number: MFCD00011248
CAS Number: 10042-76-9
UN: 1507
ADR: 5.1,III

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Titration Complexometric ≥97.5 %
Loss on drying ≤0.5 %
pH 5 to 7 (5 % at 20°C)
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR