tert-Butyl-2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

43802-0010
TS43802-0010EA 142 USD
TS43802-0010
tert-Butyl-2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate 97%
tert-Butyl-2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate
Formula: C₁₁H₁₇N₃O₂S
Melting Pt: 223.5…224.5 °C
Storage Temperature: Ambient
MDL Number: MFCD08448157
CAS Number: 365996-05-0

Order Now

Specification Test Results

Appearance Off-white to brown Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR