2-Bromoisonicotinic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Bromo-4-pyridinecarboxylic acid, 2-Bromopyridine-4-carboxylic acid

43738-0050
TS43738-0050EA 214 USD
TS43738-0050
2-Bromoisonicotinic acid 97%
2-Bromoisonicotinic acid
Formula: C₆H₄BrNO₂
MW: 202.01
Melting Pt: 212…222 °C
Storage Temperature: Ambient
MDL Number: MFCD01646069
CAS Number: 66572-56-3

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR