Sodium thiosulfate pentahydrate 99.999% (trace metals basis)
Supplier: Thermo Scientific Chemicals
AC43694-0250
AC43694-1000
TS43694-0250EA
193
USD
TS43694-0250
TS43694-1000
Sodium thiosulfate pentahydrate 99.999% (trace metals basis)
Sodium thiosulphate pentahydrate
Formula:
Na₂S₂O₃.5H₂O Melting Pt: 48.5 °C Density: 1.72 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00149186 CAS Number: 10102-17-7 |
Specification Test Results
Appearance | White Crystalline powder |
Titration Iodimetric | ≥98.5 % |
Total trace metal impurities | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...