Nickel 99.995% (trace metals basis), powder

Supplier: Thermo Scientific Chemicals
Danger

436780500
TS436780500EA 85 USD
TS436780500
Nickel 99.995% (trace metals basis), powder
Nickel
Formula: Ni
MW: 58.69 g/mol
Boiling Pt: 3007 °C (1013 hPa)
Melting Pt: 1455 °C
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
UN: 3089
ADR: 4.1,II

Order Now

Specification Test Results

Appearance Black Powder
Total trace metal impurities ≤50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR