Tetrabenzyl pyrophosphate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tetrabenzyl diphosphate

43675-0010
TS43675-0010EA 194 USD
TS43675-0010
Tetrabenzyl pyrophosphate 98%
Tetrabenzyl pyrophosphate
Formula: C₂₈H₂₈O₇P₂
MW: 538.48 g/mol
Melting Pt: 60.5…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00051941
CAS Number: 990-91-0
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 59 to 65 °C
Water ≤0.1 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR