Molecular sieve A4 (0.4 nm, 4 Å), pellets Ø 3.2mm

Supplier: Thermo Scientific Chemicals

43673-0010 436730050
TS43673-0010EA 118 USD
TS43673-0010 TS436730050
Molecular sieve A4 (0.4 nm, 4 Å), pellets Ø 3.2mm
Molecular sieve A4 (0.4 nm, 4 Å)

Order Now

Specification Test Results

Appearance Beige Cylinders
Bulk density 600 to 750 g/l
Residual water ≤2 % (550 °C, 2 h)
Water adsorption ≥20 % (50% rH, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR