Estradiol 98%

Supplier: Thermo Scientific Chemicals
Danger

43632-0050 43632-0250
TS43632-0050EA 336 USD
TS43632-0050 TS43632-0250
Estradiol 98%
Estradiol
Formula: C₁₈H₂₄O₂
Melting Pt: 176…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00003693
CAS Number: 50-28-2

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 173 to 179 °C
Water ≤3.5 %
HPLC ≥97.5 %
Specific optical rotation +76° to +83° (20 °C, 589 nm) (c=1, dioxane)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR