Glycerol-1,2-carbonate 90%

Supplier: Thermo Scientific Chemicals

43617-0250
TS43617-0250EA 186 USD
TS43617-0250
Glycerol-1,2-carbonate 90%
Glycerol-1,2-carbonate
Formula: C₄H₆O₄
Boiling Pt: 142.5 °C (1 mmHg)
Density: 1.4 g/cm³
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00085561
CAS Number: 931-40-8

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Elemental Analysis 48.8 to 59.6 % (O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR