1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
(S,S)-Et-DUPHOS-Rh, (+)-1,2-BIS((2S,5S)-2,5-DIETHYLPHOSPHOLANO)BENZENE(CYCLOOCTADIENE)RHODIUM(L)TETRAFLUOROBORATE, (+)-1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate, (+)-1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, 1,2-Bis[(2S,5S)-2,5-diethylphospholano]benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate
AC43519-1000
AC43519-2500
TS43519-1000EA
129
USD
TS43519-1000
TS43519-2500
1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate 97%
1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate
Formula:
C₃₀H₄₆BF₄P₂Rh Storage Temperature: Ambient |
MDL Number:
MFCD01862465 CAS Number: 213343-64-7 |
Specification Test Results
Appearance | Orange to red to brown Powder |
Infrared spectrum | Conforms |
NMR (1H-NMR) | Consistent with structure |
NMR (31P-NMR) | Consistent with structure |
Elemental Analysis | 52.4 to 56.8 % C |
Specific optical rotation | -40° to -50° (20°C, 589 nm) (c=1 in methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...