Butyl acetate ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Butyl ethanoate, Acetic acid butyl ester, Acetic acid n-butyl ester, n-Butyl acetate
434620025
434621000
434620010
TS434620025EA
225
USD
TS434620025
TS434621000
TS434620010
Butyl acetate ACS
Butyl acetate
Formula:
CH₃COO(CH₂)₃CH₃ MW: 116.16 g/mol Boiling Pt: 126 Melting Pt: -76 Density: 0.88 g/cm³ Flash Pt: 27 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009445 CAS Number: 123-86-4 UN: 1123 ADR: 3 III Merck Index: 13,01534 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Water | ≤0.1 % (K.F.) |
GC | ≤0.2 % (n-Butyl alcohol) |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.001 % |
Substance darkened by H2SO4 | Passes test |
Titrable acid | ≤0.0016 meq/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...