N,N,N',N'-Tetramethylethylenediamine (TEMED) 99.5%, AcroSeal™, purified by redistillation

Supplier: Thermo Scientific Chemicals
Danger

43383-1000
TS43383-1000EA 93 USD
TS43383-1000
N,N,N',N'-Tetramethylethylenediamine (TEMED) 99.5%, AcroSeal™, purified by redistillation
N,N,N',N'-Tetramethylethylenediamine (TEMED)
Formula: C₆H₁₆N₂
Boiling Pt: 120…122 °C (760 mmHg)
Melting Pt: –55 °C
Density: 0.77 g/cm³
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00008335
CAS Number: 110-18-9
UN: 2372
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6838f1f5b7d0f69dcc81a8ed247f6c178dad231c&vid=ea361367e9944e4101e38622fdd18b07756eb676&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4170 to 1.4190 (20°C, 589 nm)
GC ≥99.4 %
Color scale ≤20 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR