4-Methyl-2-pentanone 99.5% for HPLC

Supplier: Thermo Scientific Chemicals
Danger

433570010
TS433570010EA 97 USD
TS433570010
4-Methyl-2-pentanone 99.5% for HPLC
4-Methyl-2-pentanone
Formula: (CH₃)₂CHCH₂COCH₃
Boiling Pt: 117.4 °C (760 mmHg)
Melting Pt: –84 °C
Density: 0.8 g/cm³
Flash Pt: 14 °C
Storage Temperature: Ambient
MDL Number: MFCD00008938
CAS Number: 108-10-1
UN: 1245
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.05 %
GC ≥99.4 %
UV at 335 nm A: ≤1.0
Color scale ≤15 APHA
Residue after evaporation ≤0.001 %
Acidity/alkalinity ≤0.0005 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR