tert-Butylchlorodimethylsilane 50% (w/w) in toluene, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: tert-Butyldimethylsilyl Chloride, Chloro-tert-butyldimethylsilane, t-Butyldimethylchlorosilane, tert-Butyldimethylchlorosilane, tert-Butyldimethylsilyl chloride, TBDMS chloride

433111000
TS433111000EA 151 USD
TS433111000
tert-Butylchlorodimethylsilane 50% (w/w) in toluene, AcroSeal®
tert-Butylchlorodimethylsilane
Formula: C₆H₁₅ClSi
Boiling Pt: 110…126 °C (760 mmHg)
Density: 0.866 g/cm³
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00000501
CAS Number: 18162-48-6
UN: 2920
ADR: 8,I

Order Now

Specification Test Results

Appearance Clear colorless Solution
Assay 48 to 52 %
Melting point 118 to 122 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR