Methyltrimethoxysilane 97%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC43264-0010 AC43264-1000
TS43264-0010EA 95 USD
TS43264-0010 TS43264-1000
Methyltrimethoxysilane 97%, AcroSeal™
Methyltrimethoxysilane
Formula: C₄H₁₂O₃Si
Boiling Pt: 102 °C (760 mmHg)
Melting Pt: –50 °C
Density: 0.95 g/cm³
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00008342
CAS Number: 1185-55-3
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3695 to 1.3715 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR