Dichlorodiisopropylsilane 97%

Supplier: Thermo Scientific Chemicals
Danger

AC43236-0050 AC43236-0250
TS43236-0050EA 96 USD
TS43236-0050 TS43236-0250
Dichlorodiisopropylsilane 97%
Dichlorodiisopropylsilane
Formula: C₆H₁₄Cl₂Si
Boiling Pt: 168…170 °C (760 mmHg)
Density: 1.026 g/cm³
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00054895
CAS Number: 7751-38-4
UN: 2986
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4430 to 1.4450 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR