kappa-Carrageenan

Supplier: Thermo Scientific Chemicals
Warning

43159-0250 43159-1000
TS43159-0250EA 48 USD
TS43159-0250 TS43159-1000
kappa-Carrageenan
kappa-Carrageenan
Formula: C₁₂H₁₇O₁₃S
Storage Temperature: Ambient
MDL Number: MFCD00151514
CAS Number: 11114-20-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to off-white to yellow to brown Powder
Infrared spectrum Conforms
Loss on drying ≤12 % (1 g, 105°C)
Viscosity ≥10 mPa.s (0.3 % in water at 25°C)
Ash ≤40 %
Barium (Ba) ≤0.005 %
Calcium (Ca) ≤0.001 %
Lithium (Li) ≤0.001 %
Magnesium (Mg) ≤0.0002 %
Potassium (K) ≤0.03 %
Rubidium (Rb) ≤0.005 %
Sodium (Na) ≤0.02 %
Strontium (Sr) ≤0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR