Bromotrimethylsilane 98%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trimethylbromosilane, Trimethylsilyl bromide

AC43075-1000
TS43075-1000EA 157 USD
TS43075-1000
Bromotrimethylsilane 98%, AcroSeal™
Bromotrimethylsilane
Formula: C₃H₉BrSi
MW: 153.09 g/mol
Boiling Pt: 79
Melting Pt: -43
Density: 1.16 g/cm³
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00000048
CAS Number: 2857-97-8
UN: 2920
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4200 to 1.4250 (20°C, 589 nm)
Titration Argentometric ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR