6-Benzyloxy-1-boc-indole-2-boronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

43033-0010
TS43033-0010EA 222 USD
TS43033-0010
6-Benzyloxy-1-boc-indole-2-boronic acid 97%
6-Benzyloxy-1-boc-indole-2-boronic acid
Formula: C₂₀H₂₂BNO₅
Melting Pt: 102…106 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06659831
CAS Number: 850568-66-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 101 to 106 °C
HPLC ≥96.0 %
Acidity ≤0.0005 meq/g
Chloride (Cl) ≤0.001 %
Sulfate (SO4) ≤0.001 %
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Nickel (Ni) ≤0.02 ppm
Zinc (Zn) ≤0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR