Resveratrol 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: trans-3,4,5-Trihydroxystilbene, (E)-5-(4-Hydroxystyryl)benzene-1,3-diol

43007-5000
TS43007-5000EA 384 USD
TS43007-5000
Resveratrol 99%
Resveratrol
Formula: C₁₄H₁₂O₃
Melting Pt: 261…263 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00133799
CAS Number: 501-36-0

Order Now

Specification Test Results

Appearance Off-white to beige Powder
Infrared spectrum Conforms
Melting point 257 to 264 °C
Loss on drying ≤1 %
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR