Ethyl formate, extra dry 98+%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Formic acid ethyl ester

AC42924-1000
TS42924-1000EA 49 USD
TS42924-1000
Ethyl formate, extra dry 98+%, AcroSeal™
Ethyl formate
Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 54 °C (760 mmHg)
Melting Pt: –80 °C
Density: 0.917 g/cm³
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00003294
CAS Number: 109-94-4
UN: 1190
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥98.0 %
UV at 280 nm A: ≤0.015
Residue after evaporation ≤0.0003 %
Acidity ≤0.0002 meq/g
Alkalinity ≤0.0002 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR