Sodium borohydride 12% in sodium hydroxide solution 40%, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium tetrahydridoborate, Sodium tetrahydroborate

429131000
TS429131000EA 97 USD
TS429131000
Sodium borohydride 12% in sodium hydroxide solution 40%, AcroSeal®
Sodium borohydride
Formula: NaBH₄
MW: 37.83 g/mol
Boiling Pt: 130…135 °C (760 mmHg)
Melting Pt: 400 °C
Density: 1.4 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003518
CAS Number: 16940-66-2
UN: 3320
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless Liquid
Titration Iodometric 11 to 13 %
GC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR