Ethoxyacetylene 40% (w/w) in hexanes, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

429061000
TS429061000EA 645 USD
TS429061000
Ethoxyacetylene 40% (w/w) in hexanes, AcroSeal®
Ethoxyacetylene
Formula: C₄H₆O
Boiling Pt: 55…56 °C (760 mmHg)
Density: 0.718 g/cm³
Flash Pt: –28 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009247
CAS Number: 927-80-0
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear orange to brown Solution
Identification confirmed by 1H-NMR
NMR (1H-NMR) ca. 40 % w/w (typically 35 to 55 % w/w)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR