Hydrogen chloride 1 M in ethyl acetate, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

42892-1000
TS42892-1000EA 69 USD
TS42892-1000
Hydrogen chloride 1 M in ethyl acetate, AcroSeal™
Hydrogen chloride
Formula: HCl
Density: 0.9 g/cm³
Flash Pt: –3 °C
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Strength 0.95 to 1.15 M
Refractive index 1.5090 to 1.5130 (20°C, 589 nm)
GC ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR