Cyanogen bromide 3 M in dichloromethane, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

428911000
TS428911000EA 91 USD
TS428911000
Cyanogen bromide 3 M in dichloromethane, AcroSeal®
Cyanogen bromide
Formula: CBrN
Density: 1.44 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00011597
CAS Number: 506-68-3
UN: 2927
ADR: 6.1,I

Order Now

Specification Test Results

Appearance Clear colorless to yellow Solution
Strength 2.95 to 3.30 M
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR