Ethylene glycol monomethyl ether (2-methoxyethanol) for HPLC

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethylene glycol monomethyl ether

42878-0010 42878-0025
TS42878-0010EA 185 USD
TS42878-0010 TS42878-0025
Ethylene glycol monomethyl ether (2-methoxyethanol) for HPLC
Ethylene glycol monomethyl ether (2-Methoxyethanol)
Formula: C₃H₈O₂
Boiling Pt: 124 °C (760 mmHg)
Melting Pt: –85 °C
Density: 0.96 g/cm³
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00002867
CAS Number: 109-86-4
UN: 1188
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.05 %
GC ≥99.0 %
UV at 400 nm A: ≤0.01
Residue after evaporation ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR