Hydrogen chloride 5->6 N in 2-propanol, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

428601000 428608000
TS428601000EA 56 USD
TS428601000 TS428608000
Hydrogen chloride 5->6 N in 2-propanol, AcroSeal®
Hydrogen chloride
Formula: HCl
MW: 36.46
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 2920
ADR: 8 I

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Solution
Water ≤1.0 %
Titration with NaOH 5 to 6 N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR