Methyl cellulose, 400 cP (2% H₂O)

Supplier: Thermo Scientific Chemicals

Synonyms: Cellulose methyl ether

42843-0010 42843-0500 42843-2500
TS42843-0010EA 274 USD
TS42843-0010 TS42843-0500 TS42843-2500
Methyl cellulose, 400 cP (2% H₂O)
Methyl cellulose
Storage Temperature: Ambient
MDL Number: MFCD00081763
CAS Number: 9004-67-5

Order Now

Specification Test Results

Appearance White to yellowish-white to greyish-white Powder
Infrared spectrum Conforms
Loss on drying ≤5 %
Viscosity 300 to 560 cP (2 % in water at 20°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR