Tricyclohexylphosphine 1 M in toluene, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

42842-1000
TS42842-1000EA 203 USD
TS42842-1000
Tricyclohexylphosphine 1 M in toluene, AcroSeal™
Tricyclohexylphosphine
Formula: C₁₈H₃₃P
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00003853
CAS Number: 2622-14-2
UN: 1294
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Strength 0.90 to 1.10 M
Melting point 81 to 84 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR