Pivalaldehyde (trimethylacetaldehyde) 96%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Pivalaldehyde, Pivaldehyde, Trimethylacetaldehyde, 2,2-Dimethylpropionaldehyde, 2,2-Dimethylpropanal

42720-1000
TS42720-1000EA 453 USD
TS42720-1000
Pivalaldehyde (trimethylacetaldehyde) 96%, AcroSeal™
Pivalaldehyde (Trimethylacetaldehyde)
Formula: C₅H₁₀O
Boiling Pt: 74 °C (1013 hPa)
Melting Pt: 6 °C
Density: 0.775 g/cm³
Flash Pt: –15 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006962
CAS Number: 630-19-3
UN: 1989
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3770 to 1.3800 (20°C, 589 nm)
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR